Please login to your online account to display your discounted pricing

BIBR 1532, Tocris

Selective telomerase inhibitor

$233.76 - $963.39

Chemical Identifiers

Chemical Name or Material BIBR 1532
Assay Percent Range >98%
Molecular Formula C21H17NO3
Formula Weight 331.36
Synonym 2-[[(2E)-3-(2-Naphthalenyl)-1-oxo-2-butenyl1-yl]amino]benzoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

298110

 
r & d systems
RDS298110
10mg Each for $233.76

298150

 
r & d systems
RDS298150
50mg Each for $963.39
Description & Specifications

Specifications

Formula Weight 331.36
Quantity 10mg
Chemical Name or Material BIBR 1532
Assay Percent Range >98%
Molecular Formula C21H17NO3
CAS 321674-73-1
Synonym 2-[[(2E)-3-(2-Naphthalenyl)-1-oxo-2-butenyl1-yl]amino]benzoic acid
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature