Please login to your online account to display your discounted pricing

Bibenzyl 99%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: fisher scientific acros  106231000

 View more versions of this product

Catalog No. AC106231000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Crystalline Powder
Formula Weight 182.26
Quantity 100g
Color White to light yellow
TSCA TSCA
Melting Point 49° to 53°C
EINECS Number 203-096-4
RTECS Number DT4375000
CAS Max % 100.0
Packaging Glass bottle
Name Note 99%
Synonym 1, 2-Diphenylethane
Molecular Formula C14H14
Chemical Name or Material Bibenzyl
CAS 103-29-7
CAS Min % 98.5
Linear Formula C6H5CH2CH2C6H5
MDL Number MFCD00004796
Assay Percent Range 99%
Infrared Spectrum Authentic

Specifications

Chemical Name or Material Bibenzyl
Name Note 99%
CAS 103-29-7
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C14H14
Linear Formula C6H5CH2CH2C6H5
Formula Weight 182.26
MDL Number MFCD00004796
Synonym 1, 2-Diphenylethane