missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bibenzyl, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 106230250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Bibenzyl | |
103-29-7 | |
100.0 | |
White to Yellow | |
284.0°C | |
Authentic | |
Glass bottle | |
C6H5CH2CH2C6H5 | |
25 g | |
1.01 | |
1,2-diphenylethane, bibenzyl, dibenzyl, s-diphenylethane, 2-phenylethyl benzene, 1,2-dihydrostilbene, dibenzil, sym-diphenylethane, dihydrostilbene, benzene, 1,1'-1,2-ethanediyl bis | |
QWUWMCYKGHVNAV-UHFFFAOYSA-N | |
2-phenylethylbenzene | |
7647 | |
182.26 | |
Crystalline Powder |
99% | |
98.5 | |
49.0°C to 53.0°C | |
1.0100g/mL | |
129°C | |
98.5% min. (GC) | |
C14H14 | |
MFCD00004796 | |
05,598 | |
15,1202 | |
Solubility in water: practically insoluble. Other solubilities: practically insoluble in liquid ammonia, moderately soluble in alcohol, freely soluble in carbon disulfide, ether,, chloroform and amyl acetate, soluble in liquid sulfur dioxide | |
C(CC1=CC=CC=C1)C1=CC=CC=C1 | |
182.27 | |
CHEBI:34047 | |
99% |
Chemical Identifiers
103-29-7 | |
182.27 | |
QWUWMCYKGHVNAV-UHFFFAOYSA-N | |
7647 | |
2-phenylethylbenzene |
C14H14 | |
MFCD00004796 | |
1,2-diphenylethane, bibenzyl, dibenzyl, s-diphenylethane, 2-phenylethyl benzene, 1,2-dihydrostilbene, dibenzil, sym-diphenylethane, dihydrostilbene, benzene, 1,1'-1,2-ethanediyl bis | |
CHEBI:34047 | |
C(CC1=CC=CC=C1)C1=CC=CC=C1 |
Safety and Handling
EINECSNumber : 203-096-4
RTECSNumber : DT4375000
TSCA : TSCA
RUO – Research Use Only