Please login to your online account to display your discounted pricing

BI 6015, Tocris™

Hepatocyte nuclear factor 4α (HNF4α) antagonist

$231.77 - $967.24

Chemical Identifiers

Chemical Name or Material BI 6015
Assay Percent Range >98%
Molecular Formula C15H13N3O4S
Formula Weight 331.35
Synonym 2-Methyl-1-[(2-methyl-5-nitrophenyl)sulfonyl]-1H-benzimidazole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

464150

 
r & d systems
RDS464150
50mg Each for $967.24

464110

 
r & d systems
RDS464110
10mg Each for $231.77
Description & Specifications

Specifications

Formula Weight 331.35
Quantity 50mg
Chemical Name or Material BI 6015
Assay Percent Range >98%
Molecular Formula C15H13N3O4S
CAS 93987-29-2
Synonym 2-Methyl-1-[(2-methyl-5-nitrophenyl)sulfonyl]-1H-benzimidazole
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C