Please login to your online account to display your discounted pricing

BGC 20-761, Tocris

High affinity 5-HT6 antagonist

$218.74 - $915.10

Chemical Identifiers

Chemical Name or Material BGC 20-761
Assay Percent Range >98%
Molecular Formula C19H22N2O
Formula Weight 294.39
Synonym 5-Methoxy-N, N-dimethyl-2-phenyl-1H-indole-3-ethanamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

332610

 
r & d systems
332610
10mg Each for $218.74

332650

 
r & d systems
332650
50mg Each for $915.10
Description & Specifications

Specifications

Formula Weight 294.39
Quantity 10mg
Chemical Name or Material BGC 20-761
Assay Percent Range >98%
CAS 17375-63-2
Synonym 5-Methoxy-N, N-dimethyl-2-phenyl-1H-indole-3-ethanamine
Recommended Storage Store at +4°C
Molecular Formula C19H22N2O
Safety and Handling

Store at +4°C