Please login to your online account to display your discounted pricing

BF 2649 hydrochloride, Tocris™

H3 receptor inverse agonist/antagonist

$192.67 - $812.12

Chemical Identifiers

Chemical Name or Material BF 2649 hydrochloride
Assay Percent Range >99%
Molecular Formula C17H26ClNO·HCl
Formula Weight 332.31
Synonym 1-[3-[3-(4-Chlorophenyl)propoxy]propyl]-piperidine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

374310

 
r & d systems
RDS374310
10mg Each for $192.67

374350

 
r & d systems
RDS374350
50mg Each for $812.12
Description & Specifications

Specifications

Formula Weight 332.31
Quantity 10mg
Chemical Name or Material BF 2649 hydrochloride
Assay Percent Range >99%
Molecular Formula C17H26ClNO·HCl
CAS 903576-44-3
Synonym 1-[3-[3-(4-Chlorophenyl)propoxy]propyl]-piperidine hydrochloride
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature