Bevco™ Optional Caster Sets, With Mushroom Glides
Add Bevco Chair Casters to your workplace seating to provide extra mobility for workers.
Manufacturer:  Bevco™ CARD5SDUP
 View more versions of this product
Catalog No. 191302853
Description
Description
  • For use with Bevco workplace seating, including ESD and Workplace seating
  • Constructed of resilient black plastic, to accommodate up to 300 lbs.
  • Dual-wheel casters for hard floors
  • For 5-star base chairs with standard and tubular bases
Specifications
Specifications
Black
Urethane
5ST
ESD chair and stool with 5 star base
ESD hard floor caster
Documents