missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Bethanechol chloride, 98%

A cholinergic receptor agonist

$53.20 - $160.38

Chemical Identifiers

CAS 590-63-6
Molecular Formula C7H17ClN2O2
Molecular Weight (g/mol) 196.675
MDL Number MFCD00055224
InChI Key XXRMYXBSBOVVBH-UHFFFAOYSA-N
Synonym bethanechol chloride, urecholine, myocholine, besacholine, mechotane, mechothane, mecothane, myotonachol, myotonine, mictone
PubChem CID 11548
ChEBI CHEBI:3085
IUPAC Name 2-carbamoyloxypropyl(trimethyl)azanium;chloride
SMILES CC(C[N+](C)(C)C)OC(=O)N.[Cl-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6591903 View Documents Alfa Aesar™
J6591903
1g Each for $53.20
 
AAJ6591906 View Documents Alfa Aesar™
J6591906
5g Each for $160.38
 
Specifications

Chemical Identifiers

590-63-6
196.675
XXRMYXBSBOVVBH-UHFFFAOYSA-N
11548
2-carbamoyloxypropyl(trimethyl)azanium;chloride
C7H17ClN2O2
MFCD00055224
bethanechol chloride, urecholine, myocholine, besacholine, mechotane, mechothane, mecothane, myotonachol, myotonine, mictone
CHEBI:3085
CC(C[N+](C)(C)C)OC(=O)N.[Cl-]

Specifications

White
1g
C7H17ClN2O2
14,1185
XXRMYXBSBOVVBH-UHFFFAOYSA-N
2-carbamoyloxypropyl(trimethyl)azanium;chloride
11548
196.68
98%
210°C to 229°C
590-63-6
MFCD00055224
bethanechol chloride, urecholine, myocholine, besacholine, mechotane, mechothane, mecothane, myotonachol, myotonine, mictone
CC(C[N+](C)(C)C)OC(=O)N.[Cl-]
196.675
CHEBI:3085
Crystalline Powder
Bethanechol chloride
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : BR5425000

TSCA : TSCA

Recommended Storage : 2° to 8°C