Please login to your online account to display your discounted pricing

Betaxolol hydrochloride, Tocris


Selective β1 antagonist

Quantity 10mg

Manufacturer: r & d systems  RDS090610

 View more versions of this product

Catalog No. 090610

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 343.89
Synonym 1-[4-[2-(Cyclopropylmethoxy)ethyl]phenoxy]-3-isopropylamino-2-propanol hydrochloride
Chemical Name or Material Betaxolol hydrochloride
Assay Percent Range >99%
Molecular Formula C18H29NO3·HCl
CAS 63659-19-8
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material Betaxolol hydrochloride
Assay Percent Range >99%
Molecular Formula C18H29NO3·HCl
Formula Weight 343.89
Synonym 1-[4-[2-(Cyclopropylmethoxy)ethyl]phenoxy]-3-isopropylamino-2-propanol hydrochloride
Accessories

Store at Room Temperature