missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Betamethasone 21-Acetate, TCI America™

$186.39 - $635.56

Chemical Identifiers

CAS 987-24-6
Molecular Formula C24H31FO6
Molecular Weight (g/mol) 434.504
MDL Number MFCD00867462
InChI Key AKUJBENLRBOFTD-OCNKQZOYSA-N
Synonym 9alpha-Fluoro-11beta,17alpha,21-trihydroxy-16beta-methyl-1,4-pregnadiene-3,20-dione 21-Acetate
PubChem CID 45357558
IUPAC Name [2-[(9R,10S,13S,16R,17R)-9-fluoro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate
SMILES CC1CC2C3CCC4=CC(=O)C=CC4(C3(C(CC2(C1(C(=O)COC(=O)C)O)C)O)F)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B31655G View Documents TCI America
B31655G
5g Each for $635.56
 
B31651G View Documents TCI America
B31651G
1g Each for $186.39
 
Specifications

Chemical Identifiers

987-24-6
434.504
AKUJBENLRBOFTD-OCNKQZOYSA-N
45357558
CC1CC2C3CCC4=CC(=O)C=CC4(C3(C(CC2(C1(C(=O)COC(=O)C)O)C)O)F)C
C24H31FO6
MFCD00867462
9alpha-Fluoro-11beta,17alpha,21-trihydroxy-16beta-methyl-1,4-pregnadiene-3,20-dione 21-Acetate
[2-[(9R,10S,13S,16R,17R)-9-fluoro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate

Specifications

White
5g
C24H31FO6
9alpha-Fluoro-11beta,17alpha,21-trihydroxy-16beta-methyl-1,4-pregnadiene-3,20-dione 21-Acetate
CC1CC2C3CCC4=CC(=O)C=CC4(C3(C(CC2(C1(C(=O)COC(=O)C)O)C)O)F)C
434.504
434.50
≥98.0% (HPLC)
215°C
987-24-6
MFCD00867462
AKUJBENLRBOFTD-OCNKQZOYSA-N
[2-[(9R,10S,13S,16R,17R)-9-fluoro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate
45357558
Crystal-Powder at 20°C
Betamethasone 21-Acetate
Safety and Handling

Safety and Handling

EINECSNumber : (9)-0489

TSCA : No