missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Betaine monohydrate, 99+%, for analysis, Thermo Scientific Chemicals
$98.29 - $318.66
Chemical Identifiers
CAS | 590-47-6 |
---|---|
Molecular Formula | C5H13NO3 |
Molecular Weight (g/mol) | 135.16 |
MDL Number | MFCD00150010 |
InChI Key | NJZRLXNBGZBREL-UHFFFAOYSA-N |
Synonym | betaine monohydrate, 2-trimethylammonio acetate hydrate, glycine betaine hydrate, unii-tpu0itr2br, carboxymethyl trimethylammonium hydroxide, tpu0itr2br, bet hydrate, betaine, hydrate, dsstox_cid_29381, dsstox_rid_83496 |
PubChem CID | 134128033 |
IUPAC Name | carboxymethyl(trimethyl)azanium;hydroxide;hydrate |
SMILES | [OH-].C[N+](C)(C)CC(O)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC402191000
|
Thermo Scientific Chemicals
402191000 |
100 g | Glass bottle |
Each for $98.29
|
|
||||
AC402195000
|
Thermo Scientific Chemicals
402195000 |
500 g | Glass bottle |
Each for $318.66
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
590-47-6 | |
135.16 | |
NJZRLXNBGZBREL-UHFFFAOYSA-N | |
134128033 | |
[OH-].C[N+](C)(C)CC(O)=O |
C5H13NO3 | |
MFCD00150010 | |
betaine monohydrate, 2-trimethylammonio acetate hydrate, glycine betaine hydrate, unii-tpu0itr2br, carboxymethyl trimethylammonium hydroxide, tpu0itr2br, bet hydrate, betaine, hydrate, dsstox_cid_29381, dsstox_rid_83496 | |
carboxymethyl(trimethyl)azanium;hydroxide;hydrate |
Specifications
590-47-6,107-43-7 | |
100.0 | |
11% to 13.5% (140°C, 3 hrs) | |
99+% | |
C5H13NO3 | |
MFCD00150010 | |
04, 346 | |
betaine monohydrate, 2-trimethylammonio acetate hydrate, glycine betaine hydrate, unii-tpu0itr2br, carboxymethyl trimethylammonium hydroxide, tpu0itr2br, bet hydrate, betaine, hydrate, dsstox_cid_29381, dsstox_rid_83496 | |
[OH-].C[N+](C)(C)CC(O)=O | |
95% min. (c=10, 430nm) 1cm cell | |
134128033 | |
99.0 to 101.0% (on dry substance) | |
Crystalline Powder or Crystals |
99.0 | |
White | |
Authentic | |
Glass bottle | |
(CH3)3NCH2CO2H·OH | |
100 g | |
15, 1181 | |
NJZRLXNBGZBREL-UHFFFAOYSA-N | |
carboxymethyl(trimethyl)azanium;hydroxide;hydrate | |
135.16 | |
135.16 | |
Analysis | |
βine monohydrate |
Safety and Handling
EINECSNumber : 209-684-7
RUO – Research Use Only