Please login to your online account to display your discounted pricing

Betaine monohydrate, 99+%, for analysis, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: fisher scientific acros  402191000

 View more versions of this product

Catalog No. AC402191000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystals or Crystalline Powder
Formula Weight 135.16
Quantity 100g
Color White
Purity Grade For analysis
Assay 99.0 to 101.0% (on dry substance)
Beilstein 04, 346
Merck Index 15, 1181
CAS Min % 99.0
CAS Max % 100.0
Packaging Glass bottle
Assay Percent Range 99+%
Ammonium 200ppm max.
CAS 590-47-6
Synonym (Carboxymethyl)trimethylammonium hydroxide
CAS 107-43-7
Iron (Fe) 20ppm max.
Loss on Drying 11% to 13.5% (140°C, 3 hrs)
Transmittance 95% min. (c=10, 430nm) 1cm cell
Sulfate 200ppm max.
Sulfated Ash 0.1% max.
Heavy Metals (as Pb) 10ppm max.
Chlorine (Cl) 200ppm max.
Linear Formula (CH3)3NCH2CO2H·OH
Chemical Name or Material βine monohydrate
Infrared Spectrum Authentic
Molecular Formula C5H13NO3
MDL Number MFCD00150010

Specifications

Chemical Name or Material βine monohydrate
CAS 590-47-6
CAS Min % 99.0
CAS Max % 100.0
CAS 107-43-7
Purity Grade For analysis
Assay 99.0 to 101.0% (on dry substance)
Assay Percent Range 99+%
Molecular Formula C5H13NO3
Linear Formula (CH3)3NCH2CO2H·OH
Formula Weight 135.16
MDL Number MFCD00150010
Synonym (Carboxymethyl)trimethylammonium hydroxide