missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Betaine hydrochloride, 99%, extra pure, specified according to the requirements of U, Acros Organics

Manufacturer:  Fisher Scientific Acros AC164552500

 View more versions of this product

Catalog No. AC164552500



Specifications

Specifications

βine hydrochloride
0.1% max.
Plastic bottle
0.5% max.
241.0°C
0.8 to 1.2 (1:4)
98.5
99%
(CH3)3NCH2CO2H·Cl
15, 1181
Solubility in water: 647g/L (25°C). Other solubilities: soluble in ethanol,practically insoluble in chloroform and ether
C[N+](C)(C)CC(=O)O.[Cl-]
153.61
153.61
99%
A solution (1 in 20) responds to the test for chloride: Complies
Authentic
0.1% max
Colorless to White
250g
590-46-5
100.0
C5H11NO2·HCl
MFCD00011903
achylin
HOPSCVCBEOCPJZ-UHFFFAOYSA-N
carboxymethyl(trimethyl)azanium;chloride
11545
Powder or Crystals
Extra Pure

Chemical Identifiers

590-46-5
153.61
HOPSCVCBEOCPJZ-UHFFFAOYSA-N
11545
C[N+](C)(C)CC(=O)O.[Cl-]
C5H11NO2·HCl
MFCD00011903
achylin
carboxymethyl(trimethyl)azanium;chloride
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Warning

EINECSNumber : 209-683-1

SDS Documents