Please login to your online account to display your discounted pricing

Betaine, 98%, for analysis, anhydrous, ACROS Organics™


Quantity 500g
Packaging Plastic bottle

Manufacturer: fisher scientific acros  AC204245000

 View more versions of this product

Catalog No. AC204245000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Crystals or Crystalline Powder
Formula Weight 117.15
Quantity 500g
Color White
Beilstein 04, IV, 2369
Merck Index 15, 1181
Purity Grade For analysis
Assay 97.5 to 102.5%
CAS Max % 100.0
Loss on Drying 1% max. (140°C, 3 hrs)
Transmittance 95% min. (c=10, H2O, 430nm) 1cm cell
Synonym (Carboxymethyl)trimethylammonium hydroxide inner salt
Chemical Name or Material βine
Name Note Anhydrous
Sulfate 200ppm max.
Sulfated Ash 0.1% max.
MDL Number MFCD00012123
CAS 107-43-7
CAS Min % 97.5
Chlorine (Cl) 200ppm max.
Infrared Spectrum Authentic
Packaging Plastic bottle
Linear Formula (CH3)3NCH2CO2
Ammonium 200ppm max.
Heavy Metals (as Pb) 10ppm max.
Iron (Fe) 20ppm max.
Assay Percent Range 98%
Molecular Formula C5H11NO2

Specifications

Chemical Name or Material βine
Name Note Anhydrous
CAS 107-43-7
CAS Min % 97.5
CAS Max % 100.0
Purity Grade For analysis
Assay 97.5 to 102.5%
Assay Percent Range 98%
Molecular Formula C5H11NO2
Linear Formula (CH3)3NCH2CO2
Formula Weight 117.15
MDL Number MFCD00012123