missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ beta-Nicotinamide adenine dinucleotide reduced disodium salt, 98%

Manufacturer:  Alfa Aesar™ J6163803

 View more versions of this product

Catalog No. AAJ6163803



Description

Description

Applications
β-Nicotinamide adenine dinucleotide (NAD+) and β-Nicotinamide adenine dinucleotide, reduced (NADH) comprise a coenzyme redox pair (NAD+:NADH) involved in a wide range of enzyme catalyzed oxidation reduction reactions. In addition to its redox function, NAD+/NADH is a donor of ADP-ribose units in ADP-ribosylaton (ADP-ribosyltransferases; poly(ADP-ribose) polymerases ) reactions and a precursor of cyclic ADP-ribose (ADP-ribosyl cyclases). As a reagent, NADH can be used in enzyme cycling assays to amplify detection of activity of biologically relevant enzymes or metabolites present in low concentrations.

Solubility
It is soluble in water.

Notes
Store with desiccant. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature -20°C Light sensitive. Hygroscopic.
Specifications

Specifications

beta-Nicotinamide adenine dinucleotide reduced disodium salt
1g
C21H33N7Na2O14P2
5230241
It is soluble in water.
[HH].[HH].C1C=CN(C=C1C(=O)N)C2C(C(C(O2)COP(=O)(O)OP(=O)(O)OCC3C(C(C(O3)N4C=NC5=C4N=CN=C5N)O)O)O)O.[Na].[Na]
715.457
709.41
98%
White to Yellow
606-68-8
MFCD00036200
beta-nadh disodium salt
PSTAPGCXUOIFPQ-ITGWJZMWSA-N
[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoyl-4H-pyridin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl hydrogen phosphate;molecular hydrogen;sodium
131673989
Powder

Chemical Identifiers

606-68-8
715.457
PSTAPGCXUOIFPQ-ITGWJZMWSA-N
131673989
[HH].[HH].C1C=CN(C=C1C(=O)N)C2C(C(C(O2)COP(=O)(O)OP(=O)(O)OCC3C(C(C(O3)N4C=NC5=C4N=CN=C5N)O)O)O)O.[Na].[Na]
C21H33N7Na2O14P2
MFCD00036200
beta-nadh disodium salt
[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoyl-4H-pyridin-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl hydrogen phosphate;molecular hydrogen;sodium
Safety and Handling

Safety and Handling

EINECSNumber : 210-123-3

TSCA : Yes

Recommended Storage : Keep cold

Documents