missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

beta-Nicotinamide adenine dinucleotide phosphate reduced tetrasodium salt hydrate, 97%, Thermo Scientific Chemicals

$380.04 - $1319.33

Chemical Identifiers

Molecular Formula C21H26N7Na4O17P3·xH2O
MDL Number MFCD07366294
Synonym beta-NADPH tetrasodium salt tetrahydrate
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ60387MC
View Documents Promotion Details
Thermo Scientific Chemicals
J60387MC
100 mg
Each for $380.04
 
AAJ60387ME
View Documents Promotion Details
Thermo Scientific Chemicals
J60387ME
500 mg
Each for $1,319.33
 
Description

Description

beta-Nicotinamide adenine dinucleotide phosphate reduced tetrasodium salt hydrate is used as NADP+/NADPH redox pair is used in a variety of antioxidation mechanism where it protects agains reactive oxidation species accumulation. NADPH is generated in vivio by the pentose phosphate pathway.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
beta-Nicotinamide adenine dinucleotide phosphate reduced tetrasodium salt hydrate is used as NADP+/NADPH redox pair is used in a variety of antioxidation mechanism where it protects agains reactive oxidation species accumulation. NADPH is generated in vivio by the pentose phosphate pathway.

Solubility
It is soluble in 10 mM NaOH.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature -20°C Light sensitive. Hygroscopic
Specifications

Chemical Identifiers

C21H26N7Na4O17P3·xH2O
MFCD07366294
beta-NADPH tetrasodium salt tetrahydrate

Specifications

White
MFCD07366294
beta-NADPH tetrasodium salt tetrahydrate
905.42 (833.35 Anhydrous)
Powder
C21H26N7Na4O17P3·xH2O
100 mg
It is soluble in 10 mM NaOH.
≥98%
beta-Nicotinamide adenine dinucleotide phosphate reduced tetrasodium salt hydrate
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Store at -20°C

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only