Please login to your online account to display your discounted pricing

Alfa Aesar™ beta-Nicotinamide adenine dinucleotide phosphate monopotassium salt, ca 90%

$122.82 - $1,628.55

Chemical Identifiers

Chemical Name or Material beta-Nicotinamide adenine dinucleotide phosphate monopotassium salt
CAS 68141-45-7
Molecular Formula C21H27KN7O17P3
Formula Weight 781.5
MDL Number MFCD00148915
Synonym beta-NADP monopotassium salt; Triphosphopyridine nucleotide monopotassium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2445002

  SDS
alfa aesar™
B2445002
0.5g Each for $435.62

AAB2445077

  SDS
alfa aesar™
B2445077
0.1g Each for $122.82

AAB2445088

  SDS
alfa aesar™
B2445088
2.5g Each for $1,628.55
Description & Specifications

Specifications

Formula Weight 781.5
Chemical Name or Material beta-Nicotinamide adenine dinucleotide phosphate monopotassium salt
Assay ≈90%
Molecular Formula C21H27KN7O17P3
MDL Number MFCD00148915
CAS 68141-45-7
Synonym beta-NADP monopotassium salt; Triphosphopyridine nucleotide monopotassium salt
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage −30° to −10°C
Melting Point ≈175°C (dec.)
Quantity 0.5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

−30° to −10°C