missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ beta-Naphthol Violet

$50.18 - $583.39

Chemical Identifiers

CAS 7143-21-7
Molecular Formula C16H9N3Na2O9S2
Formula Weight 497.37
MDL Number MFCD00021457
Chemical Name or Material beta-Naphthol Violet
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2427909 SDS Alfa Aesar™
B2427909
10g Each for $50.18
AAB2427918 SDS Alfa Aesar™
B2427918
50g Each for $160.08
AAB2427930 SDS Alfa Aesar™
B2427930
250g Each for $583.39
Specifications

Description

Applications
Beta-Naphthol violet exhibits antiseptic properties and is used in the synthesis of certain azo dyes and antioxidants for rubber. It can also be used in the detection of the ferric ion which turns the Beta-Naphthol violet solution green.

Solubility
Soluble in water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

7143-21-7
497.37
beta-Naphthol Violet
C16H9N3Na2O9S2
MFCD00021457

Specifications

7143-21-7
497.37
Odorless
beta-Naphthol Violet
C16H9N3Na2O9S2
MFCD00021457
10g

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning