Please login to your online account to display your discounted pricing

Alfa Aesar™ beta-Galactosidase Digest Standard, Certified Mass Spec Grade


Manufacturer: alfa aesar™  J67822ODL

 View more versions of this product

Catalog No. AAJ67822ODL

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Product Type beta-Galactosidase Digest Standard
Quantity 3 vials
Storage Requirements -30°C to -10°C
CAS 9031-11-2
Format Solid
Solubility Soluble in water
Description Brown
For Use With (Application) For the standardizing, calibrating, and troubleshooting chromatographic separations. Particularly useful for LC-MS applications.