missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

beta-Estradiol, 99% (dry wt.), ca 3% water, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L0380106

 View more versions of this product

Catalog No. AAL0380106



Description

Description

β-Estradiol is the major estrogen secreted by the premenopausal ovary. It acts as an estrogenic hormone receptor (ER) agonist and activator of PI 3-kinase. Further, it is used in the treatment of infertility in women, prostate cancer and breast cancer. It is also employed as an agent of stimulating female puberty induction, as in the treatment of delayed puberty caused by hypogonadism or Turner's syndrome. In addition to this, it is used as part of the hormone replacement therapy for transgender women.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
β-Estradiol is the major estrogen secreted by the premenopausal ovary. It acts as an estrogenic hormone receptor (ER) agonist and activator of PI 3-kinase. Further, it is used in the treatment of infertility in women, prostate cancer and breast cancer. It is also employed as an agent of stimulating female puberty induction, as in the treatment of delayed puberty caused by hypogonadism or Turner′s syndrome. In addition to this, it is used as part of the hormone replacement therapy for transgender women.

Solubility
Soluble in dimethyl sulfoxide, ethanol , water, phosphate buffer saline, dimethyl formamide, acetone, dioxane and alkali hydroxides. Slightly soluble in vegetable oils.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

beta-Estradiol
175°C to 178°C
C18H24O2
5 g
14,3703
Soluble in dimethyl sulfoxide,ethanol ,water,phosphate buffer saline,dimethyl formamide,acetone,dioxane and alkali hydroxides. Slightly soluble in vegetable oils.
+79° (c=1 in Dioxane)
(8R,9S,13S,14S,17S)-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene-3,17-diol
5757
272.39
50-28-2
(dry wt.), ≈3% water
MFCD00003693
1914275
estradiol, beta-estradiol, 17beta-estradiol, oestradiol, dihydrofolliculin, estrace, ovocyclin, progynon, dihydrotheelin, dihydroxyestrin
VOXZDWNPVJITMN-ZBRFXRBCSA-N
CC12CCC3C(C1CCC2O)CCC4=C3C=CC(=C4)O
272.388
CHEBI:16469
99%

Chemical Identifiers

50-28-2
272.388
VOXZDWNPVJITMN-ZBRFXRBCSA-N
5757
(8R,9S,13S,14S,17S)-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene-3,17-diol
C18H24O2
MFCD00003693
estradiol, beta-estradiol, 17beta-estradiol, oestradiol, dihydrofolliculin, estrace, ovocyclin, progynon, dihydrotheelin, dihydroxyestrin
CHEBI:16469
CC12CCC3C(C1CCC2O)CCC4=C3C=CC(=C4)O
Safety and Handling

Safety and Handling

GHS H Statement
H350-H360
May cause cancer.
May damage fertility or the unborn child.

P201-P202-P260-P263-P264b-P270-P271-P281-P304+P340-P308+P313-P501c

H350-H361fd-H362-H373

EINECSNumber : 200-023-8

RTECSNumber : KG2975000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only