Please login to your online account to display your discounted pricing

Alfa Aesar™ beta-D-Maltose octaacetate, 98%

$39.88 - $39.88

Chemical Identifiers

Chemical Name or Material beta-D-Maltose octaacetate
CAS 22352-19-8
Molecular Formula C28H38O19
Formula Weight 678.6
MDL Number MFCD00079432
Beilstein 79279
Synonym Octa-O-acetyl-beta-D-maltose
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1298803

 
alfa aesar™
A1298803
1g Each for $39.88
Description & Specifications

Specifications

Formula Weight 678.6
Chemical Name or Material beta-D-Maltose octaacetate
Assay 98%
Molecular Formula C28H38O19
Melting Point 157° to 160°C
MDL Number MFCD00079432
CAS 22352-19-8
Beilstein 79279
Quantity 1g
Synonym Octa-O-acetyl-beta-D-maltose
Optical Rotation +65° (c=10 in Chloroform)
Recommended Storage 2° to 8°C
Safety and Handling

2° to 8°C