missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

beta-D-Allose, 97+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A16810ME

 View more versions of this product

Catalog No. AAA16810ME



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

beta-D-Allose
143°C to 146°C
MFCD00063268
1724623
14,280
C(C(C(C(C(C=O)O)O)O)O)O
180.156
180.16
2595-97-3
C6H12O6
500 mg
Hygroscopic
GZCGUPFRVQAUEE-OBOOZECYSA-N
(2R,3R,4R)-2,3,4,5,6-pentahydroxyhexanal
57449163
98%

Chemical Identifiers

2595-97-3
180.156
GZCGUPFRVQAUEE-OBOOZECYSA-N
(2R,3R,4R)-2,3,4,5,6-pentahydroxyhexanal
C6H12O6
MFCD00063268
57449163
C(C(C(C(C(C=O)O)O)O)O)O
Safety and Handling

Safety and Handling

EINECSNumber : 219-994-4

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only