missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ beta-D-Allopyranose, 97%

$85.45 - $342.35

Chemical Identifiers

CAS 7283-09-2
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
MDL Number MFCD00063268
InChI Key WQZGKKKJIJFFOK-UHFFFAOYNA-N
Synonym beta-d-allose, beta-d-allopyranose, 3 s-hydroxy-all-cis-8,11,14,17-eicosatetraenoyl-coa, 2r,3r,4r,5s,6r-6-hydroxymethyl oxane-2,3,4,5-tetrol, 1rpj, .beta.-d-allose, d +-allose, b-d-allopyranose, d-allose, beta-d-allose, rare aldohexose sugar
PubChem CID 448388
ChEBI CHEBI:40656
IUPAC Name (2R,3R,4R,5S,6R)-6-(hydroxymethyl)oxane-2,3,4,5-tetrol
SMILES C(C1C(C(C(C(O1)O)O)O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ66172MD View Documents Alfa Aesar™
J66172MD
250mg Each for $85.45
 
AAJ6617203 View Documents Alfa Aesar™
J6617203
1g Each for $342.35
 
Specifications

Chemical Identifiers

7283-09-2
180.16
WQZGKKKJIJFFOK-UHFFFAOYNA-N
448388
(2R,3R,4R,5S,6R)-6-(hydroxymethyl)oxane-2,3,4,5-tetrol
C6H12O6
MFCD00063268
beta-d-allose, beta-d-allopyranose, 3 s-hydroxy-all-cis-8,11,14,17-eicosatetraenoyl-coa, 2r,3r,4r,5s,6r-6-hydroxymethyl oxane-2,3,4,5-tetrol, 1rpj, .beta.-d-allose, d +-allose, b-d-allopyranose, d-allose, beta-d-allose, rare aldohexose sugar
CHEBI:40656
C(C1C(C(C(C(O1)O)O)O)O)O

Specifications

White
250mg
C9H10ClNO2
4-chlorophenyl dimethylcarbamate, p-chlorophenyl-n,n-dimethyl carbamate, 4-chlorophenyl n,n-dimethylcarbamate, dimethylcarbamic acid 4-chlorophenyl ester, n,n-dimethyl-4-chlorophenylcarbamate
CN(C)C(=O)OC1=CC=C(C=C1)Cl
199.63
180.16
97%
148°C to 150°C
7305-03-5
MFCD00063268
QTILATGINOFTLP-UHFFFAOYSA-N
(4-chlorophenyl) N,N-dimethylcarbamate
350036
Powder
beta-D-Allopyranose
Safety and Handling

Safety and Handling

Recommended Storage : 2° to 8°C