missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ beta-D-Allopyranose, 97%

Manufacturer:  Alfa Aesar™ J66172MD

 View more versions of this product

Catalog No. AAJ66172MD



Specifications

Specifications

beta-D-Allopyranose
148°C to 150°C
7283-09-2
MFCD00063268
QTILATGINOFTLP-UHFFFAOYSA-N
(4-chlorophenyl) N,N-dimethylcarbamate
350036
Powder
White
250mg
C9H10ClNO2
4-chlorophenyl dimethylcarbamate, 4-chlorophenyl n,n-dimethylcarbamate, dimethylcarbamic acid 4-chlorophenyl ester, n,n-dimethyl-4-chlorophenylcarbamate, p-chlorophenyl-n,n-dimethyl carbamate
CN(C)C(=O)OC1=CC=C(C=C1)Cl
199.63
180.16
97%

Chemical Identifiers

7283-09-2
199.63
QTILATGINOFTLP-UHFFFAOYSA-N
350036
CN(C)C(=O)OC1=CC=C(C=C1)Cl
C9H10ClNO2
MFCD00063268
4-chlorophenyl dimethylcarbamate, 4-chlorophenyl n,n-dimethylcarbamate, dimethylcarbamic acid 4-chlorophenyl ester, n,n-dimethyl-4-chlorophenylcarbamate, p-chlorophenyl-n,n-dimethyl carbamate
(4-chlorophenyl) N,N-dimethylcarbamate
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Keep cold

Documents