missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ beta-D-Allopyranose, 97%

Manufacturer:  Alfa Aesar™ J6617203

 View more versions of this product

Catalog No. AAJ6617203



Specifications

Specifications

beta-D-Allopyranose
148°C to 150°C
7283-09-2
MFCD00063268
WQZGKKKJIJFFOK-UHFFFAOYNA-N
(2R,3R,4R,5S,6R)-6-(hydroxymethyl)oxane-2,3,4,5-tetrol
448388
180.16
97%
White
1g
C6H12O6
beta-d-allose, beta-d-allopyranose, 3 s-hydroxy-all-cis-8,11,14,17-eicosatetraenoyl-coa, 2r,3r,4r,5s,6r-6-hydroxymethyl oxane-2,3,4,5-tetrol, 1rpj, .beta.-d-allose, d +-allose, b-d-allopyranose, d-allose, beta-d-allose, rare aldohexose sugar
C(C1C(C(C(C(O1)O)O)O)O)O
180.16
CHEBI:40656
Powder

Chemical Identifiers

7283-09-2
180.16
WQZGKKKJIJFFOK-UHFFFAOYNA-N
448388
(2R,3R,4R,5S,6R)-6-(hydroxymethyl)oxane-2,3,4,5-tetrol
C6H12O6
MFCD00063268
beta-d-allose, beta-d-allopyranose, 3 s-hydroxy-all-cis-8,11,14,17-eicosatetraenoyl-coa, 2r,3r,4r,5s,6r-6-hydroxymethyl oxane-2,3,4,5-tetrol, 1rpj, .beta.-d-allose, d +-allose, b-d-allopyranose, d-allose, beta-d-allose, rare aldohexose sugar
CHEBI:40656
C(C1C(C(C(C(O1)O)O)O)O)O
Safety and Handling

Safety and Handling

Recommended Storage : 2° to 8°C

Documents