Learn More
beta-Cyclocitral, 90+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2570018
Description
beta-Cyclocitral is used as flavoring agent, nutrient, stabilizers, surfactants and emulsifiers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationsbeta-Cyclocitral is used as flavoring agent, nutrient, stabilizers, surfactants and emulsifiers.
Solubility
Soluble in chloroform, dichloromethane, ethyl acetate and methanol. Insoluble in water.
Notes
Air sensitive. Store away from air, oxidizing agents.
Specifications
beta-Cyclocitral | |
0.944 | |
>110°C (230°F) | |
1.498 | |
50 g | |
beta-cyclocitral, b-cyclocitral, 1-cyclohexene-1-carboxaldehyde, 2,6,6-trimethyl, .beta.-cyclocitral, 2,6,6-trimethyl-1-cyclohexene-1-carboxaldehyde, unii-77y0u2x29g, cyclocitral, alpha beta-cyclocitral, 2,6,6-trimethylcyclohex-1-ene-1-carbaldehyde, 1-formyl-2,6,6-trimethyl-1-cyclohexene | |
MOQGCGNUWBPGTQ-UHFFFAOYSA-N | |
2,6,6-trimethylcyclohexene-1-carbaldehyde | |
9895 | |
152.23 |
432-25-7 | |
62°C to 63°C (3 mmHg) | |
C10H16O | |
MFCD00079078 | |
Air Sensitive | |
Soluble in chloroform,dichloromethane,ethyl acetate and methanol. Insoluble in water. | |
CC1=C(C=O)C(C)(C)CCC1 | |
152.24 | |
CHEBI:53177 | |
≥90% |
Chemical Identifiers
432-25-7 | |
152.24 | |
MOQGCGNUWBPGTQ-UHFFFAOYSA-N | |
9895 | |
2,6,6-trimethylcyclohexene-1-carbaldehyde |
C10H16O | |
MFCD00079078 | |
beta-cyclocitral, b-cyclocitral, 1-cyclohexene-1-carboxaldehyde, 2,6,6-trimethyl, .beta.-cyclocitral, 2,6,6-trimethyl-1-cyclohexene-1-carboxaldehyde, unii-77y0u2x29g, cyclocitral, alpha beta-cyclocitral, 2,6,6-trimethylcyclohex-1-ene-1-carbaldehyde, 1-formyl-2,6,6-trimethyl-1-cyclohexene | |
CHEBI:53177 | |
CC1=C(C=O)C(C)(C)CCC1 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 207-081-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only