Please login to your online account to display your discounted pricing

Alfa Aesar™ beta-Casomorphin (1-4), Amide, Bovine

For cellular and molecular biology applications

Specifications

Product Type beta-Casomorphin (1-4), amide, bovine
Cross Reactivity Bovine
Molecular Weight 521.61
Storage Requirements -30° to -10°C
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66979MA

 
alfa aesar™
J66979MA
10mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66979MB

 
alfa aesar™
J66979MB
25mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type beta-Casomorphin (1-4), amide, bovine
Cross Reactivity Bovine
Molecular Weight 521.61
Storage Requirements -30° to -10°C
Format Solid
Sequence Tyr-Pro-Phe-Pro-NH2
Regulatory Status RUO
Purification Method HPLC