missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ beta-Casomorphin (1-3)

For cellular and molecular biology applications

$231.09 - $231.09

Specifications

Product Type beta-Casomorphin (1-3)
Molecular Weight 425.48
Storage Requirements -30° to -10°C
Format Solid
Sequence Tyr-Pro-Phe
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ66782MB View Documents Alfa Aesar™
J66782MB
25mg Each for $231.09
 
Specifications

Specifications

beta-Casomorphin (1-3)
-30° to -10°C
Tyr-Pro-Phe
HPLC
425.48
Solid
RUO

For Research Use Only