Please login to your online account to display your discounted pricing

Alfa Aesar™ beta-Bromostyrene, cis + trans, 96%, stab. with 0.1% 4-tert-butylcatechol

$109.69 - $352.58

Chemical Identifiers

Chemical Name or Material beta-Bromostyrene
CAS 103-64-0
Name Note cis + trans, stabilized with 0.1% 4-tert-butylcatechol
Molecular Formula C8H7Br
Formula Weight 183.05
MDL Number MFCD00000185
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1698622

  SDS
alfa aesar™
A1698622
100g Each for $352.58

AAA1698614

  SDS
alfa aesar™
A1698614
25g Each for $109.69
Description & Specifications

Specifications

Formula Weight 183.05
Quantity 100g
Chemical Name or Material beta-Bromostyrene
Recommended Storage 2° to 8°C
Boiling Point 219° to 221°C
Molecular Formula C8H7Br
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H227
Harmful if swallowed.
Combustible liquid.
MDL Number MFCD00000185
Name Note cis + trans, stabilized with 0.1% 4-tert-butylcatechol
Assay 96%
CAS 103-64-0
Flash Point 101°C (215°F)
Health Hazard 3 GHS P Statement
P210-P280-P264-P301+P312-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point −8°C
TSCA TSCA

Applications
Beta-Bromostyrene, cis + trans is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in alcohol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under normal conditions. Keep away from strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P210-P280-P264-P301+P312-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H227
Harmful if swallowed.
Combustible liquid.

2° to 8°C