missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
β-Alanine methyl ester hydrochloride, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC371471000
Specifications
β-Alanine methyl ester hydrochloride | |
Glass bottle | |
103.0°C to 105.0°C | |
98% | |
97.5 | |
98% | |
NH2CH2CH2COOCH3·HCl | |
04, IV, 2527 | |
XPGRZDJXVKFLHQ-UHFFFAOYSA-N | |
methyl 3-aminopropanoate;hydrochloride | |
2734767 | |
Crystalline Powder |
Authentic | |
White | |
100g | |
3196-73-4 | |
100.0 | |
C4H9NO2·HCl | |
MFCD00039060 | |
3-aminopropanoic acid methyl ester hydrochloride, 3-aminopropionic acid methyl ester hydrochloride, a-ala-ome.hcl, b-alanine methyl ester hydrochloride, beta-alanine methyl ester hcl, beta-alanine methyl ester hydrochloride, h-, h-beta-ala-ome hydrochloride, h-beta-ala-ome.hcl, methyl 3-aminopropanoate hydrochloride, methyl 3-aminopropionate hydrochloride | |
COC(=O)CCN.Cl | |
139.58 | |
139.58 | |
98% |
Chemical Identifiers
3196-73-4 | |
139.58 | |
XPGRZDJXVKFLHQ-UHFFFAOYSA-N | |
2734767 | |
COC(=O)CCN.Cl |
C4H9NO2·HCl | |
MFCD00039060 | |
3-aminopropanoic acid methyl ester hydrochloride, 3-aminopropionic acid methyl ester hydrochloride, a-ala-ome.hcl, b-alanine methyl ester hydrochloride, beta-alanine methyl ester hcl, beta-alanine methyl ester hydrochloride, h-, h-beta-ala-ome hydrochloride, h-beta-ala-ome.hcl, methyl 3-aminopropanoate hydrochloride, methyl 3-aminopropionate hydrochloride | |
methyl 3-aminopropanoate;hydrochloride |