Please login to your online account to display your discounted pricing

Alfa Aesar™ [beta-Ala&superscript_8;] Neurokinin A (4-10)


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J66848EXD

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ66848EXD

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Format Powder
Molecular Weight 780.94
Storage Requirements -30° to -10°C
Quantity 0.5mg
Regulatory Status RUO
Product Type [beta-Ala8] Neurokinin A (4-10)
Sequence Asp-Ser-Phe-Val-β-Ala-Leu-Met-NH2
Purification Method HPLC