Please login to your online account to display your discounted pricing

Alfa Aesar™ [beta-Ala&superscript_8;] Neurokinin A (4-10)

For cellular and molecular biology applications

Specifications

Product Type [beta-Ala8] Neurokinin A (4-10)
Molecular Weight 780.94
Storage Requirements -30° to -10°C
Format Powder
Sequence Asp-Ser-Phe-Val-β-Ala-Leu-Met-NH2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66848EXD

 
alfa aesar™
J66848EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66848MCR

 
alfa aesar™
J66848MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type [beta-Ala8] Neurokinin A (4-10)
Molecular Weight 780.94
Storage Requirements -30° to -10°C
Format Powder
Sequence Asp-Ser-Phe-Val-β-Ala-Leu-Met-NH2
Regulatory Status RUO
Purification Method HPLC