Please login to your online account to display your discounted pricing

Alfa Aesar™ beta,2-Dinitrostyrene, 97%

$105.58 - $105.58

Chemical Identifiers

Chemical Name or Material beta,2-Dinitrostyrene
CAS 3156-39-6
Molecular Formula C8H6N2O4
Formula Weight 194.15
MDL Number MFCD00052066
Beilstein 1964093
Synonym 2-Nitro-1-(2-nitrophenyl)ethene; 2-Nitro-beta-nitrostyrene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1289103

 
alfa aesar™
L1289103
1g Each for $105.58
Description & Specifications

Specifications

Formula Weight 194.15
Chemical Name or Material beta,2-Dinitrostyrene
Assay 97%
Molecular Formula C8H6N2O4
MDL Number MFCD00052066
CAS 3156-39-6
Synonym 2-Nitro-1-(2-nitrophenyl)ethene; 2-Nitro-beta-nitrostyrene
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 104° to 109°C
Beilstein 1964093
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.