SHOWA™ Value Master™ Natural Rubber Gloves
Resist animal fats, caustics, acids, oils, salts, alcohols, and detergents
Manufacturer:  SHOWA™ VML09
 View more versions of this product
Catalog No. 11-395-23C
Specifications Description & Specifications
Specifications
12 in.
Yellow
30.5cm
manufacturing, aircraft assembly, janitorial, plant maintenance, food processing, poultry processing, sanitization clean up, household cleaning
Tractor tread
Flock
Rubber, natural
18mil
21 CFR 170-199, CE
Rolled
Large