Please login to your online account to display your discounted pricing
SHOWA™ Value Master™ Natural Rubber Gloves
Resist animal fats, caustics, acids, oils, salts, alcohols, and detergents
Manufacturer: showa™  VMUPM08
 View more versions of this product
Catalog No. 11-395-18B
  • / Case of 144 Pair
Description & Specifications
Specifications
Certifications 21 CFR 170-199, CE
Glove Cuff Style Rolled
For Use With (Application) manufacturing, aircraft assembly, janitorial, plant maintenance, food processing, poultry processing, sanitization clean up, household cleaning
Material Natural Rubber
Exterior Finish Tractor tread
Length (Metric) 30.5cm
Color Blue
Size Medium
Thickness 18mil
Length (English) 12 in.
Lining Flock