SHOWA™ Nu Fangle™ and NuFangle Plus Gloves
Cool and comfortable
Manufacturer: SHOWA™ 7610**
Description
Alternative to cotton and lightweight leather gloves.
- Nitrile-impregnated coating
- Full impregnated or ventilated back
- Cotton knit liner
- Puncture- and snag-resistant
- Full-cut, slip-on design
- Sanitized™
- Color-coded for easy size identification
Specifications
Fully Impregnated | |
Textured | |
assembly, maintenance, inspection, warehousing, shipping and receiving | |
Cotton | |
Navy |
Full | |
Slip-on | |
Nitrile | |
Medium |