Please login to your online account to display your discounted pricing

SHOWA™ Nitri-Flex™ Kevlar™ Gloves

Flexible nitrile-dipped gloves with Kevlar™ liner provide comfort with durability

$108.87 - $108.87

Specifications

Color Green
Color Yellow
Cut Resistance Rating Level 2
Material Kevlar™
For Use With (Application) Shearing, Metal fabrication and assembly, general construction, facilities maintenance, warehouse and shipping jobs
View More Specs
Products
Catalog Number Mfr. No. Length (English) Length (Metric) Size Price Quantity    

19-050-303D

 
showa™
490010
10 in. 25.5cm X-Large Dozen of 12 Pair for $108.87
Case of 6 Dozen for $653.22
Description & Specifications

Specifications

Color Green
Color Yellow
Cut Resistance Rating Level 2
Material Kevlar™
For Use With (Application) Shearing, Metal fabrication and assembly, general construction, facilities maintenance, warehouse and shipping jobs
Glove Cuff Style Knit
Hand Hand-specific
Weight Medium weight

Suitable for a variety of industrial applications; permit wearer the dexterity to handle small parts.

  • Nitrile coating covers fingers and palm, offering excellent grip in wet, dry, or oily applications
  • Resists cuts, snags and puncture, providing abrasion resistance that outwears cotton and leather gloves five to one
  • Uncoated back allows heat to dissipate through seamless liner
  • Sizes are stamped on gloves; knitwrists are color coded for easy sorting
  • Hand specific; launderable without bleach