missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
SHOWA™ Viton™ and Viton II Gloves
Provide highest chemical-resistance barrier to aromatic hydrocarbons such as benzene, toluene, or xylene.
Manufacturer: SHOWA™ 89211
Description
Provide highest chemical-resistance barrier to aromatic hydrocarbons such as benzene, toluene, or xylene.
- Unsupported, unlined gloves made of Viton over butyl are heavy-duty and flexible
- Smooth finish and rolled cuffs
- Thicker Viton II gloves also provde a barrier to most chlorinated solvents and aliphatic hydrocarbons
Specifications
Smooth | |
Provides high temperature and resistance to fuels used in the aerospace industry | |
Butyl,Viton™ | |
12 in. | |
CE |
Black | |
Rolled | |
11 | |
30.5 cm | |
12 mil |