SHOWA™ Viton™ and Viton II Gloves
Provide highest chemical-resistance barrier to aromatic hydrocarbons such as benzene, toluene, or xylene.
Manufacturer:  SHOWA™ 89210
 View more versions of this product
Catalog No. 19050070D
Description
Description
Provide highest chemical-resistance barrier to aromatic hydrocarbons such as benzene, toluene, or xylene.
  • Unsupported, unlined gloves made of Viton over butyl are heavy-duty and flexible
  • Smooth finish and rolled cuffs
  • Thicker Viton II gloves also provde a barrier to most chlorinated solvents and aliphatic hydrocarbons
Specifications
Specifications
Black
Rolled
10
30.5 cm
12 mil
Provides high temperature and resistance to fuels used in the aerospace industry
Butyl,Viton™
12 in.
CE
Smooth