Please login to your online account to display your discounted pricing
SHOWA™ Glove™ Nitri-Solve™ Unlined Nitrile Gloves
Manufacturer: showa™  717L09
 View more versions of this product
Catalog No. 11-395-29C
  • / Pair
  • / Case of 144 Pair
Description & Specifications
Specifications
Certifications 21 CFR 170-199, NFPA 1992, CFIA, CE
Glove Cuff Style Straight
For Use With (Application) manufacturing, chemical processing, Petrochemicals, food processing, aerospace, automobile degreasing, automotive assembly, painting applications, machine applications using cutting Oils
Length (Metric) 33cm
Size Large
Coating Material Bisque
Color Light green
Length (English) 13 in.
Material Nitrile
Thickness 11mil