Please login to your online account to display your discounted pricing
SHOWA™ Glove™ Nitri-Solve™ Unlined Nitrile Gloves
Manufacturer: showa™  717S07
 View more versions of this product
Catalog No. 11-395-29A
  • / Pair
  • / Case of 144 Pair
Description & Specifications
Specifications
Length (English) 13 in.
Length (Metric) 33cm
Color Light Green
Thickness (English) 11mil
Certifications 21 CFR 170-199, NFPA 1992, CFIA, CE
For Use With (Application) manufacturing, chemical processing, Petrochemicals, food processing, aerospace, automobile degreasing, automotive assembly, painting applications, machine applications using cutting Oils
Glove Cuff Style Straight
Size Small
Material Nitrile
Coating Material Bisque