Please login to your online account to display your discounted pricing
SHOWA™ SHOWA™ Industrial Unlined Nitrile Green Gloves, 12-pack
Wear SHOWA™ Industrial Unlined Nitrile Green Gloves for applications in manufacturing, chemical processing, Petrochemicals, food processing, aerospace, automobile degreasing, automotive assembly, painting applications and more.
Manufacturer: showa™  747XXL11
 View more versions of this product
Catalog No. 11-395-32C
  • / Pair
  • / Case of 72 Pair
Description & Specifications
Specifications
Coating Material Bisque
Glove Cuff Style Straight
For Use With (Application) manufacturing, chemical processing, Petrochemicals, food processing, aerospace, automobile degreasing, automotive assembly, painting applications, machine applications using cutting Oils
Material Nitrile
Size 2X-Large
Thickness (English) 22mil
Certifications 21 CFR 170-199, NFPA 1992, CFIA, CE
Color Light Green
Length (English) 19 in.
Length (Metric) 48.26cm
  • Cat III
  • 12-pack
  • Case-hardened finish enhances chemical and abrasion resistance
  • Easy to pull on and off
  • TAA compliant