SHOWA™ Glove™ Nitri-Solve™ Flock-Lined Nitrile Gloves
Nitri Solve 730 offers a light-weight chemical resistant glove with premium nitrile formulation for handling light solvents
Manufacturer:  SHOWA™ 730XXL11
 View more versions of this product
Catalog No. 11-395-33E
Specifications Description & Specifications
Specifications
Bisque
manufacturing, food processing, hazmat splash protection, laboratories, janitorial, paint applications, fuel handling and refineries
Nitrile
15mil
Green
33cm
Straight
Flock
11
21 CFR 170-199, NFPA 1992, CFIA, CE
13 in.