Please login to your online account to display your discounted pricing

SHOWA™ Glove™ Nitri-Flex™ Lite and Nitri-Flex Plus Nitrile-Coated Gloves

$4.13 - $54.74

Specifications

Cleaning Procedure Launder at 40°C using a neutral Detergent
Coated Area Palm
Color Light green, green
Cut Resistance Rating Level 1
Material Nitrile
View More Specs
Products
Catalog Number Mfr. No. Exterior Finish Liner Material Length (English) Size Price Quantity    

19-050-283A

 
showa™
450007
Smooth Nylon 8.5 in. 7 Dozen of 12 Pair for $47.59
Case of 6 Dozen for $285.54

29-635-1292

 
showa™
4500M08
Smooth Nylon 9 in. 8 Pair for $4.13
Case of 72 Pair for $268.56

19-050-283C

 
showa™
4500L09*
Smooth Nylon 9 in. 9 Dozen of 12 Pair for $49.31
Case of 6 Dozen for $295.86

19-050-283D

 
showa™
4500XL10
Smooth Nylon 10 in. 10 Dozen of 12 Pair for $54.74
Case of 6 Dozen for $328.44
Description & Specifications

Specifications

Cleaning Procedure Launder at 40°C using a neutral Detergent
Coated Area Palm
Color Light green, green
Cut Resistance Rating Level 1
Material Nitrile
For Use With (Application) small parts assembly, inspection, shipping and receiving, light fabrication, material handling, automotive, packaging
Glove Cuff Style Knit

Nitri-Flex™ Lite

  • Flat-dipped nitrile, seamless nylon liner
  • Smooth palm
  • Light green with green dip
Nitri-Flex™ Plus
  • Flat dipped nitrile, seamless cotton liner
  • Provides superior absorption of perspiration
  • Textured palm provides excellent wet, dry, or oily grip