missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

SHOWA™ Nitri-Flex™ Lite and Nitri-Flex Plus Nitrile-Coated Gloves

Ideal for small parts assembly, inspection, shipping and receiving, light fabrication, material handling, automotive, packaging

Manufacturer:  SHOWA™ 4500M08

Catalog No. 29-635-1292




Description

Description

Nitri-Flex™ Lite

  • Flat-dipped nitrile, seamless nylon liner
  • Smooth palm
  • Light green with green dip
Nitri-Flex™ Plus
  • Flat dipped nitrile, seamless cotton liner
  • Provides superior absorption of perspiration
  • Textured palm provides excellent wet, dry, or oily grip

Specifications

Specifications

Green,Light Green
Launder at 40°C using a neutral detergent
Knit
Nitrile
9 in.
Level 1
Palm
small parts assembly, inspection, shipping and receiving, light fabrication, material handling, automotive, packaging
Nylon
8
23 cm
Smooth
SDS
Documents