missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

SHOWA™ 707D Chemical-Protection Nitrile Gloves

The 707D chemical-protection gloves combine the best of disposable and chemical-resistant technology. This hybrid results in the ideal solution for jobs requiring contact with chemical hazards, optimum comfort, and tactility.

Supplier:  SHOWA™ 707XXL11

 View more versions of this product

Catalog No. 29-664-134




Description

Description

  • Form-fitting
  • Easy donning and doffing
  • Chemical-resistant
  • Oil-resistant
  • Water-resistant
  • Abrasion-resistant
  • Hydrocarbon-resistant
  • Impermeable
  • Biodegradable
  • Ergonomic
  • Rolled Cuff
  • 100% Nitrile
  • Chlorinated
  • Embossed grip
  • Unlined
  • Eco Best Technology™ (EBT)
  • Industries: Chemical, Food, Janitorial, Laboratory, Mining, Pharmaceutical
Specifications

Specifications

21 CFR 170-199, ANSI/ISEA 105-2005
Yes
2X-Large
manufacturing, food processing, laboratories, janitorial, paint applications, poultry processing, light duty maintenance and clean up
Rough
30.5 cm
Blue
Nitrile
9 mil
Rolled
12 in.
SDS
Documents

Documents

Promotions

Promotions