SHOWA™ Nitri-Dex Nitrile Gloves
Manufacturer:  SHOWA™ 707XL10
 View more versions of this product
Catalog No. 29-664-133
Description
Description
Specifications
Specifications
9 mil
manufacturing, food processing, laboratories, janitorial, paint applications, poultry processing, light duty maintenance and clean up
X-Large
12 in.
21 CFR 170-199, CE, ASTM, ANSI/ISEA 105-2005, EN 388, EN 374-3-2003, CE
Rolled
Nitrile
Blue
30.5 cm