SHOWA™ Nitri-Dex™ Flock-lined Gloves
Manufacturer:  SHOWA™ 707FLM08
 View more versions of this product
Catalog No. 19-150-1292
Specifications
Specifications
21 CFR 170-199, ANSI/ISEA 105-2005, CE, EN 388-2003, EN 374-3-2003, ASTM
Rolled
Cotton Flock
Medium
Blue
30.5cm
Tractor tread
manufacturing, laboratories, paint applications, fuel handling, food processing, janitorial, agricultural chemicals, refineries, light duty maintenance and clean up, paint applications, poultry processing
Nitrile
11mil
12 in.
Documents