missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
SHOWA™ Nitri-Dex™ Flock-Lined Nitrile Gloves
Unsupported, flock-lined nitrile coated gloves feature a lasting grip and provide protection from oils, hydrocarbons, grease and abrasion.
Supplier: SHOWA™ 707FL11
Description
Unsupported, flock-lined nitrile coated gloves feature a lasting grip and provide protection from oils, hydrocarbons, grease and abrasion.
- Ideal for jobs requiring comfort, tactility, and contact with chemical hazards
- Flocked cotton liner ensures easy donning and doffing
- Rolled cuff prevents debris from entering
- Lightweight nitrile material helps reduce hand fatigue
Specifications
ANSI 1 | |
CE | |
Blue | |
Nitrile | |
9 mil | |
EN 388:2003 1001 | |
JKL | |
Manufacturing, laboratories, paint applications, fuel handling, food processing, janitorial, agricultural chemicals, refineries, light duty maintenance and clean up, paint applications, poultry processing | |
Tractor tread | |
Lined | |
SHOWA™ 707FL | |
30.5 cm |
21 CFR 170-199 | |
No | |
Yes | |
2X-Large | |
0.23 mm | |
No | |
Bacteria, Fungi only | |
Gauntlet | |
Cotton Flock | |
707FL-11 | |
12 in. |