SHOWA™ Chem Master™ Neoprene/Natural Rubber Gloves
Self-flushing, tractor-tread grip pattern improves gripping effectiveness by encouraging fluid run-off
Manufacturer:  SHOWA™ CHML09
 View more versions of this product
Catalog No. 11-394-114C
Alert:
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Specifications Description & Specifications
Specifications
9
Powder-free
Non-sterile
30.5cm
21 CFR 170-199
Straight
Flock
Latex, natural rubber,Neoprene
Black,Blue
26mil
12 in.
chemical plants, automotive and battery manufacturing, pesticide manufacturing and plant maintenance
Tractor tread